Product Name
Lithium zirconate −80 mesh
Product Number
400920
Product Brand
ALDRICH
CAS Number
12031-83-3
Molecular Weight
153.10
Test | Specification |
---|---|
APPEARANCE | OFF-WHITE POWDER |
X-RAY DIFFRACTION | CONFORMS TO STANDARD PATTERN. |
AND/OR | |
ICP ASSAY | CONFIRMS LITHIUM AND ZIRCONIUM COMPONENTS. |
LOSS ON IGNITION | TYPICALLY <2% |
(1100 DEGREES C TO CONSTANT WEIGHT) | |
PARTICLE SIZE | -80 MESH |
APPROVED MAY 30, 2000 JSB |