Product Name
(1S,2S)-trans-1,2-Cyclohexanediol 99%
Product Number
421804
Product Brand
ALDRICH
CAS Number
57794-08-8
Molecular Weight
116.16
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER, CRYSTALS, CRYSTALLINE POWDER |
AND/OR CHUNKS | |
IDENTITY | CONFORMS TO STRUCTURE BY IR OR HNMR SPECTRA. |
OPTICAL ROTATION | +39(+/-1) DEGREES (C=1.6%, H2O) |
ENANTIOMERIC EXCESS | 98.5% (MINIMUM) |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED OCTOBER 31, 2007 RJM |