Product Name
Orcinol 97%
Product Number
447420
Product Brand
ALDRICH
CAS Number
504-15-4
Molecular Weight
124.14
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Conforms to Requirement |
Powder, Crystals and/or Chunks | |
Infrared spectrum | Conforms to Structure |
GC (area %) | ≥96.5 % |