Product Name
(1S, 2S)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride 97%
Product Number
684031
Product Brand
ALDRICH
CAS Number
1052707-07-9
Molecular Weight
375.21
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Conforms |
Powder, Crystals, Crystalline Powder and/or Chunks | |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 96.5 - 103.5 % |
Optical Rotation | -88.0 - -80.0 |
Degrees C=1%, H2O | |
Purity (HPLC) | ≥96.5 % |