Product Name
4-Formylphenylboronic acid MIDA ester 96%
Product Number
697494
Product Brand
ALDRICH
CAS Number
Molecular Weight
261.04
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN SOLID, POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 52.7% TO 57.7% |
NITROGEN 5.1% TO 5.6% | |
APPROVED MARCH 14, 2008 DJP |