Product Name
1,1′,4′,1′′-Terphenyl-4-thiol 97%
Product Number
708488
Product Brand
ALDRICH
CAS Number
90589-98-3
Molecular Weight
262.37
Test | Specification |
---|---|
Appearance (Color) | Yellow to Orange |
Appearance (Form) | Conforms to Requirements |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Carbon | 79.51 - 85.28 % |
Sulfur | 11.79 - 12.64 % |
Purity (GC) | ≥96.5 % |