Product Name
Palladium on barium sulfate 5% Pd basis
Product Number
76022
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Faint Brown to Dark Brown |
APPEARANCE (FORM) | Powder |
ASSAY (ATOMIC ABSORPT.) | 4.8 - 5.2 % PD |
ICP ANALYSIS | CONFIRMS PD, BA, S COMPONENT |