Product Name
D-(−)-Isoascorbic acid 98%
Product Number
856061
Product Brand
ALDRICH
CAS Number
89-65-6
Molecular Weight
176.12
Test | Specification |
---|---|
APPEARANCE | WHITE CRYSTALLINE POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5% - 102.5% (WITH NAOH) |
OPTICAL ROTATION | -16.8(+/- 1.5) DEGREES (C=2%, H2O) |
GAS LIQUID | 97.5% (MINIMUM) |
SOLUBILITY | 50 MG/ML H2O; CLEAR, COLORLESS SOLUTION |
REVISED OCTOBER 11, 2000 JSB |