Product Name
Cyclobutanecarbonyl chloride 98%
Product Number
C95706
Product Brand
ALDRICH
CAS Number
5006-22-4
Molecular Weight
118.56
Test | Specification |
---|---|
Appearance (Color) | Colorless |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 97,5 - 102,5 % |
GC (area %) | ≥97,5 % |