Product Name
2-Thiophenecarboxylic acid hydrazide ≥98%
Product Number
T1388
Product Brand
ALDRICH
CAS Number
2361-27-5
Molecular Weight
142.18
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow |
APPEARANCE (FORM) | Powder to Crystals |
PURITY (GC AREA %) | ≥ 98 % |
SOLUBILITY (COLOR) | Colorless to Light Yellow |
SOLUBILITY (TURBIDITY) | Clear to Very Slightly Hazy |
SOLUBILITY (METHOD) | 200 mg plus 4ml of ETHANOL |
CARBON CONTENT | 41.7 - 42.7 % |
NITROGEN CONTENT | 19.3 - 20.1 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 72 MONTHS |