Product Name
m-Tolylacetic acid 99%
Product Number
T38091
Product Brand
ALDRICH
CAS Number
621-36-3
Molecular Weight
150.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (T) NAOH 0.1M | 98.5 - 101.5 % |
PURITY (GC AREA %) | ≥ 98.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |