Product Name
Diethylene glycol analytical standard
Product Number
03128
Product Brand
SIAL
CAS Number
111-46-6
Molecular Weight
106.12
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Viscous Liquid |
COLOR (IN APHA) | ≤ 25 APHA |
PURITY (GC AREA %) | ≥ 99.5 % |
DENSITY D20/4 | 1.115 - 1.117 |
REFRACTIVE INDEX N20/D | 1.446 - 1.448 |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |