Product Name
Abberior® STAR 635P, streptavidin
Product Number
04683
Product Brand
SIAL
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Report Result |
APPEARANCE (FORM) | Solid or Semi-Solid or Liquid |
EMISSION MAXIMUM | 651 ± 10 nm |
REMARKS ON LUMINESCENCE | EXC. 638 nm in Water |
DYE-TO-PROTEIN RATIO (MOLAR) | Report Result |
SDS GEL ELECTROPHORESIS | CORRESPONDS TO REQUIREMENTS |
REM. ON GELELECTROPHORESIS | unconjugated dye < 5% of total fluorescence |