Product Name
Fluorescein sodium salt analytical standard
Product Number
30181
Product Brand
SIAL
CAS Number
518-47-8
Molecular Weight
376.27
Test | Specification |
---|---|
APPEARANCE (COLOR) | Light Red to Very Dark Red |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 97.5 % |
WATER | ≤ 15 % |
CARBON CONTENT | 63.84% (Theory, anhydrous) |
HYDROGEN CONTENT | 2.68% (Theory, anhydrous) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
ASSAY (ICP) | 12.22% (Theory, Disodium salt, anhydrous) |
REMARKS ON ICP | Na positive |
EXPIRY PERIOD | 36 MONTHS |