Product Name
Meloxicam-d3 VETRANAL®, analytical standard
Product Number
34109
Product Brand
SIAL
CAS Number
942047-63-4
Molecular Weight
354.42
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 99.0 % |
MELTING POINT | 244 - 250 C |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
ISOTOPIC ENRICHMENT(D) | ≥ 98.0 % |
RESIDUAL SOLVENTS | ≤ 1.0 % |
EXPIRY PERIOD | 60 MONTHS |