Product Name
Carbofuran-3-hydroxy PESTANAL®, analytical standard
Product Number
37896
Product Brand
SIAL
CAS Number
16655-82-6
Molecular Weight
237.25
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
MELTING POINT | 136 - 142 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |