Product Name
1-Phenyldodecane analytical standard
Product Number
44178
Product Brand
SIAL
CAS Number
123-01-3
Molecular Weight
246.43
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.5 % |
REFRACTIVE INDEX N20/D | 1.481 - 1.483 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |