Product Name
(R)-(+)-α-Methylbenzyl isocyanate derivatization grade (chiral), LiChropur™, ≥99.0%
Product Number
77968
Product Brand
SIAL
CAS Number
33375-06-3
Molecular Weight
147.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Very Faint Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
ENANTIOMERIC RATIO | ≥ 99.5 : 0.5 % |
SPECIFIC ROTATION (20/D) | 10.5 ± 0.5 Degrees |
CONCENTRATION | NEAT |
DENSITY D20/4 | 1.040 - 1.050 |
REFRACTIVE INDEX N20/D | 1.512 - 1.514 |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 28 MONTHS |