Product Name
Sodium bromate ≥99%
Product Number
02151
Product Brand
SIGALD
CAS Number
7789-38-0
Molecular Weight
150.89
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Crystals |
REDOX TITRATION | ≥ 99 % |
TRACE ANALYSIS (ADDITIONAL TESTS) | total impurities ≤ 0.005% heavy metals (as Pb) |
IRON (ICP) | ≤ 50 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 500 mg/kg |
BROMIDE (BR) | ≤ 500 mg/kg |
RECOMMENDED RETEST PERIOD | 54 MONTHS |