Product Name
Pyridine ReagentPlus®, ≥99%
Product Number
320498
Product Brand
SIGALD
CAS Number
110-86-1
Molecular Weight
79.10
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Faint Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |