Product Name
Anthracene suitable for scintillation, ≥99.0% (GC)
Product Number
10580
Product Brand
SIGMA
CAS Number
120-12-7
Molecular Weight
178.23
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 99.0 % |
MELTING POINT | 214 - 217 C |
CARBON CONTENT | 94.34 % (Theory) |
HYDROGEN CONTENT | 5.66 % (Theory) |
EMISSION MAXIMUM | 401.0 ± 2.0 nm |
REMARKS ON LUMINESCENCE | Exc.: 251 nm in acetonitrile |
APPLICATION TEST | sublimation residue ≤ 0.1 % |