Product Name
Cholesterol Esterase from porcine pancreas lyophilized, powder, white, ~35 U/mg
Product Number
26745
Product Brand
SIGMA
CAS Number
9026-00-0
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 5 MG/ML 0.1 M PHOSPHATE PH 7.0 |
ENZYMATIC ACTIVITY | 28.0 - 50.0 U/mg |
RECOMMENDED RETEST PERIOD | 24 MONTHS |