Product Name
D-Glyceric acid sodium salt ≥95.0% (TLC)
Product Number
61786
Product Brand
SIGMA
CAS Number
6000-40-4
Molecular Weight
106.08 (free acid basis)
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (TLC AREA %) | ≥ 95.0 % |
SPECIFIC ROTATION (20/D) | 18.5 ± 3.0 Degrees |
CONCENTRATION | C = 1 in water |
CARBON CONTENT | 28.14 % (theory) |
HYDROGEN CONTENT | 4.13 % (theory) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 48 MONTHS |