Product Name
Ammonium sulfate 2M solution
Product Number
76399
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
DENSITY D20/4 | 1.129 - 1.135 |
REFRACTIVE INDEX N20/D | 1.368 - 1.372 |
PH | 5.8 - 6.2 |