Product Name
Fluorescent IEF-Marker pI 7.6 solution 1 mg/mL in 10 mM HCl
Product Number
89952
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Faint Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPCE AREA %) | ≥ 80.0 % |
EMISSION MAXIMUM | 495 ± 5 nm |
REMARKS ON LUMINESCENCE | EX. 385 NM IN 0.1 M PHOSPHATE PH 7.6 |
HPCE TEST | PI CORRESPONDS |
RECOMMENDED RETEST PERIOD | 42 MONTHS |