Product Name
Amiloride hydrochloride hydrate ≥98% (HPLC), powder
Product Number
A7410
Product Brand
SIGMA
CAS Number
2016-88-8
Molecular Weight
266.09 (anhydrous basis)
Test | Specification |
---|---|
APPEARANCE | LIGHT YELLOW TO YELLOW WITH A LIGHT GREEN CAST POWDER |
SOLUBILITY | CLEAR YELLOW TO GREEN SOLUTION AT 50MG/ML IN WATER |
WATER BY KARL FISCHER | NMT 16.9% (3MOL/MOL) |
UV-VIS SPECTRUM | LAMBDA MAX AT 361-363, 285-286 AND 212-214 NM IN WATER |
PURITY BY THIN LAYER CHROMATOGRAPHY | NLT 98% |
RECOMMENDED RETEST | 3 YEARS |
DOCUMENT # A7410/06/01/04/2 |