Product Name
BME Amino Acids Solution 50× Without L-glutamine, sterile-filtered, BioReagent, suitable for cell culture
Product Number
B6766
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | CLEAR SOLUTION |
PH TEST AT RT | 1.5 - 2.0 |
STERILITY BY USP | STERILE |
ENDOTOXIN ASSAY | NMT 1 EU/ML |
AMINO ACIDS BY HPLC | CONSISTENT WITH FORMULA |
CELL CULTURE TEST | PASS |
B6766:2/98-3 |