Product Name
Z-L-Lys-ONp hydrochloride
Product Number
C3637
Product Brand
SIGMA
CAS Number
2179-15-9
Molecular Weight
437.87
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS TO FAINT YELLOW SOLUTION AT 50 MG/ML IN METHANOL |
CARBON | 53.5 - 56.2% |
NITROGEN | 9.3 - 9.9% |
SPECIFIC ROTATION | -24.6 +/- 1 DEG (C = 1 IN DIMETHYL FORMAMIDE AT 24 DEG C) |
PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 98% |
DOCUMENT # | C3637/05/12/08/1 |