Product Name
L-Lysine p-nitroanilide dihydrobromide
Product Number
L7002
Product Brand
SIGMA
CAS Number
40492-96-4
Molecular Weight
428.12
Test | Specification |
---|---|
APPEARANCE | OFF- WHITE POWDER |
SOLUBILITY | CLEAR TO SLIGHTLY HAZY FAINT YELLOW SOLUTION AT 50 MG/ML IN WATER |
IR SPECTRUM | CONSISTENT WITH STRUCTURE |
WATER BY KARL FISCHER | REPORT RESULT |
CARBON | 32.8 - 34.5% * |
NITROGEN | 12.7 - 13.4% * |
SPECIFIC ROTATION | +60 +/- 2 DEG (C=1 IN WATER AT 25 DEG) |
PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 98% |
RECOMMENDED RETEST | 4 YEARS |