Product Name
Anti-PNPLA3 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401851
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Tissue lysate) | Antibody concentration: 1 - 5 ug/ml. Specific band of ~45.77 kDa using human kidney lysate. |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~52.80 kDa using PNPLA3 transfected 293T lysate. |
Detection Antibody | Suitable as a detection antibody in sandwich ELISA assay. |