Product Name
Anti-6-His antibody produced in rabbit affinity isolated antibody
Product Number
SAB4301134
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | Affinity Purified |
Western Blotting | 1:500-1:2000 |