Product Name
Anti-Phospho-Smad3-S423/S425 antibody produced in rabbit
Product Number
SAB5701792
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
Immunofluorescence | 1:50 - 1:200 |