Product Name
ECE-1 Substrate ≥95% (HPLC)
Product Number
SCP0131
Product Brand
SIGMA
CAS Number
Molecular Weight
1388.44
Test | Specification |
---|---|
Form | lyophilized powder |
Appearance | Off-white to yellow |
Peptide Content | > or = to 50% |
Purity by HPLC | > or = to 95% |