Product Name
Monoclonal Anti-G3BP antibody produced in mouse clone 2F3, purified immunoglobulin, buffered aqueous solution
Product Number
WH0010146M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~52 kDa using A-431 lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 1 ug/ml using human lymphoma tissue. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged G3BP is approximately 1ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~36 kDa using immunogen protein lysate. |