Product Name
Diclobutrazol reference material
Product Number
04357
Product Brand
SUPELCO
CAS Number
75736-33-3
Molecular Weight
328.24
Test | Specification |
---|---|
Assay (QNMR)* | Report certified value and uncertainty (acc. to ISO17025) |
APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 94.0 % |
EXPIRY PERIOD | 36 MONTHS |