Product Name
trans-2-Decenoyl-L-carnitine-(N-methyl-d3) analytical standard
Product Number
14811
Product Brand
SUPELCO
CAS Number
Molecular Weight
316.45
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
PURITY (HPLC AREA %) | ≥ 95.0 % |
IDENTIFICATION (LC-MS) | Identity Confirmed |
REMARKS ON LC-MS | Degree of Labelling ≥ 99 % |
SPECIFIC ROTATION (20/D) | -18.0 - -12.0 Degrees |
CONCENTRATION | C=0.1 in Water |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |