Skip to Content
Merck
1-(1-isobutoxyethoxy)-2-methylpropane AldrichCPR

1-(1-isobutoxyethoxy)-2-methylpropane

1-(1-isobutoxyethoxy)-2-methylpropane AldrichCPR
Empirical Formula (Hill Notation):
C10H22O2
CAS Number:
Molecular Weight:
174.28

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing