Skip to Content
Merck
(16a)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione Pharmaceutical Analytical Impurity (PAI)

(16a)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione

(16a)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
(16α)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione, (2-((8S,9S,10R,13S,14S,16R,17S)-16,17-Dihydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl acetate)
Empirical Formula (Hill Notation):
C23H28O7
CAS Number:
Molecular Weight:
416.46

Products

Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade