Skip to Content
Merck
1-Cyclohexylethanone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-Cyclohexylethanone

1-Cyclohexylethanone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Acetylcyclohexane, Cyclohexyl methyl ketone, Hexahydroacetophenone
Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade