Skip to Content
Merck
vibrant-m

1H,1H,2H,2H-Perfluorododecanesulfonic acid

vibrant-m
Synonym(s):
10:2 Fluorotelomer sulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododecane-1-sulfonic acid
Empirical Formula (Hill Notation):
C12H5F21O3S
CAS Number:
Molecular Weight:
628.20

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Application
Reference Material Type
Special Grade