
Atorvastatin Methyl Ester

Synonym(s):
Atorvastatin methyl ester, (methyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate), Methyl (βR,δR)-2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoate
Empirical Formula (Hill Notation):
C34H37FN2O5
CAS Number:
Molecular Weight:
572.67
Products
Feature
Formula Weight
Manufacturer
Agency
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing