Skip to Content
Merck
Peptone from casein and other animal proteins suitable for microbiology

Casein Peptone suitable for biotechnology and microbiology

Peptone from casein and other animal proteins suitable for microbiology
Synonym(s):
Peptone from animal tissue
CAS Number:

Products

Product Category
Biological Source
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application