
Chlorothiophene Aldehyde

Synonym(s):
5-Chloro-2-thiophenecarboxaldehyde
Empirical Formula (Hill Notation):
C5H3ClOS
CAS Number:
Molecular Weight:
146.59
Products
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Agency
Format
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing