Skip to Content
Merck
Cyclam United States Pharmacopeia (USP) Reference Standard

Cyclam

Cyclam United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,4,8,11-Tetraazacyclotetradecane, Cyclam
Empirical Formula (Hill Notation):
C10H24N4
CAS Number:
Molecular Weight:
200.32

Products

Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing