Skip to Content
Merck
Deferasirox for system suitability CRS European Pharmacopoeia (EP) Reference Standard

Deferasirox for system suitability CRS

Deferasirox for system suitability CRS European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Deferasirox, 4-[3,5-bis(2-Hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoic acid
Empirical Formula (Hill Notation):
C21H15N3O4
CAS Number:
Molecular Weight:
373.36

Products

Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1