Skip to Content
Merck
vibrant-m

Diclofenac diethylamine

vibrant-m
Synonym(s):
Diclofenac diethylamine salt, N-Ethylethanamine 2-[(2,6-dichlorophenyl)amino]benzeneacetate
Empirical Formula (Hill Notation):
C14H11Cl2NO2· C4H11N
CAS Number:
Molecular Weight:
369.29

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing