Skip to Content
Merck
E. coli O157:H7 (non-toxigenic) WDCM 00014 Vitroids™ 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

E. coli O157:H7 (non-toxigenic) WDCM 00014 Vitroids

E. coli O157:H7 (non-toxigenic) WDCM 00014 Vitroids™ 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Escherichia coli O157:H7 (non-toxigenic) WDCM 00014 Vitroids

Products

Brand
Sustainability
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade