
L-Glucono-delta-lactone

Synonym(s):
L-Glucono-delta-lactone, ((3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one)
Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Products
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing