Skip to Content
Merck
Ligustrazine phyproof® Reference Substance

Ligustrazine

Ligustrazine phyproof® Reference Substance
Synonym(s):
2,3,5,6-Tetramethylpyrazine
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing