Skip to Content
Merck
N-Azidoacetylglucosamine-tetraacylated 95%

N-Azidoacetylglucosamine-tetraacylated

N-Azidoacetylglucosamine-tetraacylated 95%
Empirical Formula (Hill Notation):
C16H22N4O10
CAS Number:
Molecular Weight:
430.37

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing